Anti-FCER2 Mouse Monoclonal Antibody [clone: [EBVCS2]]

51-340
10076-780EA 540.97 USD
10076-780
Anti-FCER2 Mouse Monoclonal Antibody [clone: [EBVCS2]]
FCER2 antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2, 150 mM NaCl, 0.09% sodium azide, 0.2% BSA

Store FCER2 antibody at 4 °C. As with all antibodies avoid freeze/thaw cycles

Type: Primary
Antigen: FCER2
Clonality: Monoclonal
Clone: [EBVCS2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR