Anti-CD1B Mouse Monoclonal Antibody [clone: [SN13/K5-1B8]]

51-336
10076-772EA 674.53 USD
10076-772
Anti-CD1B Mouse Monoclonal Antibody [clone: [SN13/K5-1B8]]
Antibodies
CD1B antibody can be used in ELISA starting at 1:50 - 1:100, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence.

PBS, pH 7.2, \<=0.09% sodium azide

Store CD1B antibody at 4 °C. As with all antibodies avoid freeze/thaw cycles

Type: Primary
Antigen: CD1B
Clonality: Monoclonal
Clone: [SN13/K5-1B8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR