Anti-ERBB3 Mouse Monoclonal Antibody [clone: [3F10F6]]

51-318
10076-738EA 674.53 USD
10076-738
Anti-ERBB3 Mouse Monoclonal Antibody [clone: [3F10F6]]
Antibodies
ERBB3 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ERBB3
Clonality: Monoclonal
Clone: [3F10F6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR