Anti-CK7 Mouse Monoclonal Antibody [clone: [5D12]]

Supplier: Prosci

51-311
10076-724EA 561.89 USD
10076-724
Anti-CK7 Mouse Monoclonal Antibody [clone: [5D12]]
Antibodies
Cytoke antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: [5D12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR