Anti-APOA4 Mouse Monoclonal Antibody [clone: [1D4C11]]

51-306
10076-714EA 697.97 USD
10076-714
Anti-APOA4 Mouse Monoclonal Antibody [clone: [1D4C11]]
Antibodies
APOA4 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: APOA4
Clonality: Monoclonal
Clone: [1D4C11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR