Anti-ALCAM Mouse Monoclonal Antibody [clone: [3F8B12]]

51-304
10076-710EA 698.16 USD
10076-710
Anti-ALCAM Mouse Monoclonal Antibody [clone: [3F8B12]]
Antibodies
ALCAM antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ALCAM
Clonality: Monoclonal
Clone: [3F8B12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR