Anti-BRAF Mouse Monoclonal Antibody [clone: [1H12]]

51-300
10074-694EA 674.53 USD
10074-694
Anti-BRAF Mouse Monoclonal Antibody [clone: [1H12]]
Antibodies
  • BRAF antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 μg/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BRAF
Clonality: Monoclonal
Clone: [1H12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR