Anti-CA1 Mouse Monoclonal Antibody [clone: [9D6D7]]

51-298
10079-490EA 684.46 USD
10079-490
Anti-CA1 Mouse Monoclonal Antibody [clone: [9D6D7]]
Antibodies
Carbonic Anhydrase I antibody can be used in Western Blot starting at 1:2000, and immunohistochemistry starting at 5 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CA1
Clonality: Monoclonal
Clone: [9D6D7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR