Anti-ST6GAL1 Mouse Monoclonal Antibody

Supplier: Prosci

51-287
10079-468EA 624.31 USD InStock
10079-468
Anti-ST6GAL1 Mouse Monoclonal Antibody
Antibodies
CDw75 antibody can be used in Western Blot starting at 1:100 - 1:1000, and immunohistochemistry starting at 1:50.

10mM PBS, pH 7.4, with 0.2% BSA and 0.09% Sodium Azide

CDw75 antibody is stable for 24 months when stored at 2-8 °C.

Type: Primary
Antigen: ST6GAL1
Clonality: Monoclonal
Clone: [LN1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR