Anti-CCNB1 Mouse Monoclonal Antibody

Supplier: Prosci

51-285
10079-464EA 561.89 USD InStock
10079-464
Anti-CCNB1 Mouse Monoclonal Antibody
Antibodies
Cyclin B1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS 50% glycerol, 0.09% azide

Cyclin B1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: CCNB1
Clonality: Monoclonal
Clone: [V63]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Guinea pig, Hamster, Human, Mouse, Pig, Rabbit, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR