Anti-HLA-E Mouse Monoclonal Antibody [clone: [MEM-E/07]]

Supplier: Prosci

51-282
10079-458EA 582.9 USD
10079-458
Anti-HLA-E Mouse Monoclonal Antibody [clone: [MEM-E/07]]
Antibodies
HLA-E antibody can be used in ELISA, Western Blot starting at 1 - 2 ?g/mL, immunohistochemistry starting at 1:50, and immunoprecipitation

PBS, 15 mM sodium azide, approx., pH 7.4.

HLA-E antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: HLA-E
Clonality: Monoclonal
Clone: [MEM-E/07]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR