Anti-HLA-E Mouse Monoclonal Antibody [clone: [MEM-E/06]]

Supplier: Prosci

51-281
10079-456EA 582.74 USD
10079-456
Anti-HLA-E Mouse Monoclonal Antibody [clone: [MEM-E/06]]
Antibodies
HLA-E antibody can be used in Western Blot, and immunohistochemistry starting at 1:50.

Tris buffered saline (TBS), 15 mM sodium azide, approx., pH 8.0.

HLA-E antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: HLA-E
Clonality: Monoclonal
Clone: [MEM-E/06]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR