Anti-IGHA1 Mouse Monoclonal Antibody [clone: [AD3]]

Supplier: Prosci

51-280
10079-454EA 582.9 USD
10079-454
Anti-IGHA1 Mouse Monoclonal Antibody [clone: [AD3]]
Antibodies
IgA antibody can be used in immunohistochemistry starting at 5 ?g/mL.

20 mM Tris/HCl, 15 mM sodium azide, approx., pH 7.4.

IgA antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: IGHA1
Clonality: Monoclonal
Clone: [AD3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR