Anti-CK18 Mouse Monoclonal Antibody [clone: [DA-7]]

51-277
10074-688EA 540.97 USD
10074-688
Anti-CK18 Mouse Monoclonal Antibody [clone: [DA-7]]
Cytoke antibody can be used in Western Blot starting at 1 - 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, 15 mM sodium azide, approx., pH 7.4.

Cytoke antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: KRT18
Clonality: Monoclonal
Clone: [DA-7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR