Anti-CK18 Mouse Monoclonal Antibody [clone: [C-04]]

51-275
10074-684EA 540.97 USD
10074-684
Anti-CK18 Mouse Monoclonal Antibody [clone: [C-04]]
Cytoke antibody can be used in immunohistochemistry starting at 10 ?g/mL.

PBS, 15 mM sodium azide, approx., pH 7.4.

Cytoke antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: KRT18
Clonality: Monoclonal
Clone: [C-04]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mammal
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR