Anti-LPCAT1 Rabbit Polyclonal Antibody

51-268
10074-670EA 684.46 USD
10074-670
Anti-LPCAT1 Rabbit Polyclonal Antibody
Antibodies
LPCAT1 antibody can be used in Western Blot starting at 1 - 4 ?g/mL, immunohistochemistry starting at 10 ?g/mL, immunocytochemistry starting at 4 ?g/mL, immunoprecipitation, and flow cytometry starting at 1 - 3 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LPCAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR