Anti-C5AR1 Rabbit Polyclonal Antibody

51-266
10074-666EA 698.16 USD
10074-666
Anti-C5AR1 Rabbit Polyclonal Antibody
Antibodies
C5AR1 antibody can be used in Western Blot starting at 1 ?g/mL, immunohistochemistry in frozen tissues starting at 1 ?g/mL, and immunocytochemistry.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: C5AR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Gorilla, Human, Monkey, Orangutan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR