Anti-SGK2 Rabbit Polyclonal Antibody

51-258
10074-654EA 674.53 USD
10074-654
Anti-SGK2 Rabbit Polyclonal Antibody
Antibodies
SGK2 antibody can be used in immunohistochemistry starting at 5 - 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SGK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Bovine, Gorilla, Human, Orangutan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR