Anti-NR1D2 Rabbit Polyclonal Antibody

51-257
10076-708EA 698.16 USD
10076-708
Anti-NR1D2 Rabbit Polyclonal Antibody
Antibodies
NR1D2 antibody can be used in immunohistochemistry starting at 20 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NR1D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Bovine, Chicken, Dog, Gorilla, Horse, Human, Monkey, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR