Anti-ELTD1 Rabbit Polyclonal Antibody

Supplier: Prosci

51-254
10076-702EA 561.89 USD InStock
10076-702
Anti-ELTD1 Rabbit Polyclonal Antibody
Antibodies
ELTD1 antibody can be used in immunohistochemistry starting at 2.5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ELTD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR