Anti-TAAR5 Rabbit Polyclonal Antibody

Supplier: Prosci

51-252
10076-698EA 582.9 USD
10076-698
Anti-TAAR5 Rabbit Polyclonal Antibody
Antibodies
TAAR5 antibody can be used in immunohistochemistry starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TAAR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR