Anti-MRGPRF Rabbit Polyclonal Antibody

51-249
10076-692EA 674.53 USD
10076-692
Anti-MRGPRF Rabbit Polyclonal Antibody
Antibodies
MRGPRF antibody can be used in immunohistochemistry starting at 20 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MRGPRF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR