Anti-GPR132 Rabbit Polyclonal Antibody

51-248
10076-690EA 674.53 USD
10076-690
Anti-GPR132 Rabbit Polyclonal Antibody
Antibodies
GPR132 antibody can be used in immunohistochemistry starting at 10 - 20 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR132
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR