Anti-MUC16 Mouse Monoclonal Antibody [clone: [X306]]

51-138
10074-650EA 540.97 USD
10074-650
Anti-MUC16 Mouse Monoclonal Antibody [clone: [X306]]
Mucin 16 monoclonal antibody was concluded by the International Society of Oncology and BioMarkers (ISOBM) to contain two major antigenic domains. Recognizes the epitope specificity group A, similar to the OC125 antibody. Not suitable for conjugation to HRP

Mucin 16 antibody can be used in ELISA, and immunohistochemistry starting at 5 ?g/mL.

Ion Exchange Chromatography

Tris buffered saline, 0.09% sodium azide.

Store Mucin 16 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: MUC16
Clonality: Monoclonal
Clone: [X306]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR