Anti-LRRC7 Rabbit Polyclonal Antibody

Supplier: Prosci

51-134
10074-642EA 582.9 USD
10074-642
Anti-LRRC7 Rabbit Polyclonal Antibody
Antibodies
LRRC7 antibody can be used in Western Blot, immunohistochemistry starting at 20 ?g/mL, immunofluorescence, and immunoprecipitation

PBS, 0.01% sodium azide, 50% glycerol.

Store LRRC7 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: LRRC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR