Anti-RNF125 Rabbit Polyclonal Antibody

51-133
10074-640EA 491.23 USD
10074-640
Anti-RNF125 Rabbit Polyclonal Antibody
RNF125 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 ?g/mL, and immunofluorescence.

Sera, 50% glycerol, 0.025% sodium azide.

Store RNF125 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: RNF125
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chimpanzee, Dog, Horse, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR