Anti-PPP3R1 Rabbit Polyclonal Antibody

51-131
10074-636EA 491.23 USD
10074-636
Anti-PPP3R1 Rabbit Polyclonal Antibody
PPP3R1 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 1.25 ?g/mL.

0.1M Tris-glycine, 0.15M NaCl, 0.05% sodium azide, before the addition of glycerol to 30%.

Store PPP3R1 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: PPP3R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Drosophila, Human, Mouse, Rat, Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR