Anti-DISC1 Rabbit Polyclonal Antibody

Supplier: Prosci

51-126
10074-626EA 582.9 USD
10074-626
Anti-DISC1 Rabbit Polyclonal Antibody
Antibodies
DISC1 antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

PBS, 0.01% sodium azide, 50% glycerol.

Store DISC1 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: DISC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR