Membrane-spanning 4-domains, Subfamily A, Member 1 (MS4A1) (CD20) Mouse anti-Human Monoclonal (MEM-269) Antibody. Reacts with CD20 (Bp35), a 33-37 kDa non-glycosylated membrane receptor with four transmembrane domains, expressed on B lymphocytes (it is lost on plasma cells), follicular dendritic cells, and at low levels on peripheral blood T lymphocytes.
MS4A1 antibody can be used in Western Blot starting at 1 - 2 ?g/mL, immunohistochemistry starting at 5 ?g/mL, immunoprecipitation, and immunofluorescence.
Hydrophobic Interaction Chromatography (HIC)
PBS, 15 mM sodium azide, approx., pH 7.4.
Store MS4A1 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.
Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: [MEM-269]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human