Anti-PAG1 Mouse Monoclonal Antibody [clone: [MEM-255]]

51-122
10074-618EA 684.46 USD
10074-618
Anti-PAG1 Mouse Monoclonal Antibody [clone: [MEM-255]]
Antibodies
Recongizes an epitope (aa 235-280) of Csk-binding protein (Cbp) located in the cytoplasmic domain, also known as protein associated with glycosphingolipid-enriched microdomains (PAG).

PAG1 antibody can be used in ELISA starting at 1:100000, Western Blot starting at 1:1000 - 1:10000, and immunohistochemistry starting at 5 ?g/mL.

Protein A Column

PBS, 15 mM sodium azide, approx., pH 7.4.

Store PAG1 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: PAG1
Clonality: Monoclonal
Clone: [MEM-255]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR