Anti-S100 A4 Rabbit Polyclonal Antibody

Supplier: Prosci

51-116
10079-446EA 582.9 USD
10079-446
Anti-S100 A4 Rabbit Polyclonal Antibody
Antibodies
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.

S100A4 antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, immunocytochemistry, and immunoprecipitation

10 mM PBS, pH 7.4, BSA, sodium azide.

Store S100A4 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: S100A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR