Anti-APC Rabbit Polyclonal Antibody

51-115
10079-444EA 540.97 USD
10079-444
Anti-APC Rabbit Polyclonal Antibody
APC is associated with structural components of intracellular junctions, including conductin, and it competes with E-cadherin for binding to specific internal regions of both - and -catenin. Like APC, APC2 contains SAMP domains, which are required for conductin binding. Both APC and APC2 regulate the formation of active -catenin-Tcf complexes.

APC antibody can be used in ELISA, Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

10 mM PBS, pH 7.4, BSA, sodium azide.

Store APC antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: APC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR