Anti-EPB41L1 Mouse Monoclonal Antibody

51-111
10079-438EA 491.23 USD
10079-438
Anti-EPB41L1 Mouse Monoclonal Antibody
This antibody recognizes serine, threonine, and tyrosine phosphorylated proteins as determined by ELISA using phosphorylated peptides. Reactivity has also been shown with phosphorylated proteins in western blots.

Phospho-Ser/Thr/Tyr antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 5 ?g/mL.

Protein G Column

PBS, pH 7.6, 1% BSA, with less than 0.1% sodium azide

Store Phospho-Ser/Thr/Tyr antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: EPB41L1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR