Anti-GZMB Rabbit Polyclonal Antibody

Supplier: Prosci
51-110
10079-436EA 865 USD
10079-436
Anti-GZMB Rabbit Polyclonal Antibody
Antibodies
Granzyme A and granzyme B are serine proteases that mediate apoptotic signaling in cytotoxic T lymphocytes (CTL) and in natural killer (NK) cells. Both granzyme A and granzyme B are synthesized as inactive proenzymes, and they are stored within cytolytic granules and released by effector cells during degradation. This antibody should be useful for the localization of granzyme B-containing lytic granules and for the characterization of activated CTLs or NK cells.

GZMB antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 10 ?g/mL.

10 mM PBS, pH 7.4, BSA, sodium azide.

Store GZMB antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: GZMB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR