Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LY75 Rabbit Polyclonal Antibody

Supplier: Prosci

51-092
10076-658EA 467.84 USD InStock
10076-658
Anti-LY75 Rabbit Polyclonal Antibody
Antibodies
LY75 antibody can be used in ELISA, Western Blot starting at 1 - 5 μg/mL, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

Purified

20 mM Tris-HCl, pH 8.0, 20 mg/mL BSA, 0.05% sodium azide

Store LY75 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type:
Antigen: LY75
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty