Anti-S100 A1 Rabbit Polyclonal Antibody

Supplier: Prosci

51-091
10076-656EA 582.9 USD
10076-656
Anti-S100 A1 Rabbit Polyclonal Antibody
Antibodies
S100A4 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

20 mM Tris-HCl, pH 8.0, 20 mg/mL BSA, 0.05% sodium azide

Store S100A4 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: S100A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR