Anti-CYAT1 Rabbit Polyclonal Antibody

51-081
10076-638EA 684.46 USD
10076-638
Anti-CYAT1 Rabbit Polyclonal Antibody
Antibodies
Immunoglobulin Lambda Light Chain antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

20 mM Tris-HCl, pH 8.0, 20 mg/mL BSA, 0.05% sodium azide

Store Immunoglobulin Lambda Light Chain antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: CYAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR