Anti-ACTB Rabbit Polyclonal Antibody

51-080
10076-636EA 674.53 USD
10076-636
Anti-ACTB Rabbit Polyclonal Antibody
Antibodies
  • Beta Actin antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 30 μg/mL.

Purified

20 mM Tris-HCl, pH 8.0, 20 mg/mL BSA, 0.05% sodium azide

Store Beta Actin antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: ACTB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR