Anti-ICAM1 Mouse Monoclonal Antibody

51-076
10076-628EA 750.52 USD
10076-628
Anti-ICAM1 Mouse Monoclonal Antibody
Antibodies
ICAM1 antibody can be used in Western Blot starting at 0.5 - 1 ?g/mL, and immunohistochemistry starting at 5 ?g/mL. Flow Cytometry 1:50-1:100 Immunohistochemistry (frozen)- 1:50-1:100, Immunohistochemistry (FFPE), Immunoprecipitation. Flow Cytometry: Use 10ul of the suggested working dilution to label 10e6 cells in 100ul. Immunohistochemistry Recommended positive control tissue: Human Tonsil. Recognizes the human CD54 cell surface antigen, a 90kD glycoprotein also known as ICAM-1. CD54 is expressed by many cells following activation by inflammatory mediators. Clone 3H1547 blocks CD54 function. Removal of sodium azide is recommended prior to use in functional assays.

PBS, pH 7.4, 0.09% sodium azide

Store ICAM1 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR