Anti-VWF Mouse Monoclonal Antibody

51-075
10076-626EA 491.23 USD
10076-626
Anti-VWF Mouse Monoclonal Antibody
VWF antibody can be used in immunohistochemistry starting at 20 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide, 0.2% BSA, 50% glycerol

VWF antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: VWF
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR