Anti-DISC1 Rabbit Polyclonal Antibody

51-068
10076-614EA 674.53 USD
10076-614
Anti-DISC1 Rabbit Polyclonal Antibody
Antibodies
DISC1 antibody can be used in immunohistochemistry starting at 1.3 ?g/mL.

PBS, 0.01% sodium azide, 50% glycerol.

Store DISC1 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: DISC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR