Anti-SCN2B Rabbit Polyclonal Antibody

Supplier: Prosci

51-062
10076-604EA 582.9 USD
10076-604
Anti-SCN2B Rabbit Polyclonal Antibody
Antibodies
SCN2B antibody can be used in immunohistochemistry starting at 10 ?g/mL.

PBS, 0.01% sodium azide, 50% glycerol.

Store SCN2B antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: SCN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR