Anti-SCNN1G Rabbit Polyclonal Antibody

Supplier: Prosci

51-059
10076-598EA 582.9 USD
10076-598
Anti-SCNN1G Rabbit Polyclonal Antibody
Antibodies
SCNN1G antibody can be used in immunohistochemistry starting at 5 ?g/mL.

PBS, 0.01% sodium azide, 50% glycerol.

Store SCNN1G antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: SCNN1G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR