Anti-CD40 Mouse Monoclonal Antibody [clone: [LOB7/6]]

Supplier: Prosci

51-054
10076-588EA 582.9 USD
10076-588
Anti-CD40 Mouse Monoclonal Antibody [clone: [LOB7/6]]
Antibodies
CD40 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, and immunohistochemistry starting at 5 ?g/mL. This product requires protein digestion pre-treatment of paraffin embedded sections prior to staining eg. pronase or trypsin. IHC Control: Human Tonsil or Spleen

Tris buffered saline, 0.09% sodium azide.

Store CD40 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles. Store undiluted.

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: [LOB7/6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR