Anti-VIL1 Mouse Monoclonal Antibody

Supplier: Prosci

51-050
10076-582EA 582.9 USD
10076-582
Anti-VIL1 Mouse Monoclonal Antibody
Antibodies
Villin 1 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

10 mM PBS, pH 7.4, 0.2% BSA, 15 mM sodium azide.

For long term storage of Villin 1 antibody add glycerol (40-50%) and store at -20 °C. For short term store at 4 °C.

Type: Primary
Antigen: VIL1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR