Anti-IGHA1 Mouse Monoclonal Antibody [clone: [Mc24-2E11]]

51-048
10079-410EA 684.46 USD
10079-410
Anti-IGHA1 Mouse Monoclonal Antibody [clone: [Mc24-2E11]]
Antibodies
Immunoglobulin A antibody can be used in immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IGHA1
Clonality: Monoclonal
Clone: [Mc24-2E11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR