Anti-FGA Sheep Polyclonal Antibody

Supplier: Prosci
51-047
10079-408EA 785.47 USD
10079-408
Anti-FGA Sheep Polyclonal Antibody
Antibodies
Recognises human fibrinogen, a 340kD glycoprotein which plays an important role in the coagulation process. This antibody shows minimal cross-reactivity with related serum proteins.

Fibrinogen antibody can be used in ELISA, and immunohistochemistry starting at 5 ?g/mL.

Protein G Column

PBS, pH 7.4, 0.09% sodium azide.

Store Fibrinogen antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles. Store undiluted.

Type: Primary
Antigen: FGA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR