Anti-MMP9 Mouse Monoclonal Antibody

51-046
10079-406EA 674.53 USD
10079-406
Anti-MMP9 Mouse Monoclonal Antibody
Antibodies
MMP9 antibody can be used in Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.09% sodium azide, 0.2% BSA, 40% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MMP9
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR