Anti-SMAD3 Mouse Monoclonal Antibody [clone: [AF9F7]]

51-041
10079-396EA 698.16 USD
10079-396
Anti-SMAD3 Mouse Monoclonal Antibody [clone: [AF9F7]]
Antibodies
SMAD3 antibody can be used in immunohistochemistry starting at 10 ?g/mL.

HEPES with 0.15M NaCl, 0.01% BSA, 0.03% sodium azide, and 50% glycerol

Store SMAD3 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: SMAD3
Clonality: Monoclonal
Clone: [AF9F7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR