Anti-CTSK Rabbit Polyclonal Antibody

Supplier: Prosci

51-032
10079-378EA 582.9 USD
10079-378
Anti-CTSK Rabbit Polyclonal Antibody
Antibodies
Cathepsin K antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2, 30% glycerol, 0.5% BSA, 0.01% Thimerosal.

Store Cathepsin K antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: CTSK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR